missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)ethanolamine, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 303170250

 View more versions of this product

Catalog No. AC303170250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-(tert-Butoxycarbonyl)ethanolamine
26690-80-2
100.0
>110°C
97.5% min. (GC)
C7H15NO3
HOCH2CH2NHCO2C(CH3)3
25mL
n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NCCO
161.2
161.2
98%
97.5
1.0420g/mL
Authentic
Glass bottle
1.4485 to 1.4505
MFCD00056657
1.042
GPTXCAZYUMDUMN-UHFFFAOYSA-N
tert-butyl N-(2-hydroxyethyl)carbamate
2733206
98%

Chemical Identifiers

26690-80-2
161.2
GPTXCAZYUMDUMN-UHFFFAOYSA-N
2733206
CC(C)(C)OC(=O)NCCO
C7H15NO3
MFCD00056657
n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
tert-butyl N-(2-hydroxyethyl)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only