missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)-D-prolinol, TCI America™

$21.39 - $64.17

Chemical Identifiers

CAS 83435-58-9
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD00040580
InChI Key BFFLLBPMZCIGRM-MRVPVSSYSA-N
Synonym boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate, n-tert-butoxycarbonyl-d-prolinol
PubChem CID 688279
IUPAC Name tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30761G View Documents TCI America
B30761G
1g Each for $21.39
 
B30765G View Documents TCI America
B30765G
5g Each for $64.17
 
Specifications

Chemical Identifiers

83435-58-9
201.266
BFFLLBPMZCIGRM-MRVPVSSYSA-N
688279
CC(C)(C)OC(=O)N1CCCC1CO
C10H19NO3
MFCD00040580
boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate, n-tert-butoxycarbonyl-d-prolinol
tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Specifications

White-Yellow
1g
C10H19NO3
boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate, n-tert-butoxycarbonyl-d-prolinol
CC(C)(C)OC(=O)N1CCCC1CO
201.266
201.27
≥98.0% (GC)
62°C
83435-58-9
MFCD00040580
BFFLLBPMZCIGRM-MRVPVSSYSA-N
tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
688279
Crystal-Powder at 20°C
N-(tert-Butoxycarbonyl)-D-prolinol
Safety and Handling

Safety and Handling

TSCA : No