missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)-D-proline Methyl Ester, TCI America™

$64.17 - $210.84

Chemical Identifiers

CAS 73323-65-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00190828
InChI Key WVDGSSCWFMSRHN-MRVPVSSYSA-N
Synonym boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
PubChem CID 1277667
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B47871G View Documents TCI America
B47871G
1g Each for $64.17
 
B47875G View Documents TCI America
B47875G
5g Each for $210.84
 
Specifications

Chemical Identifiers

73323-65-6
229.276
WVDGSSCWFMSRHN-MRVPVSSYSA-N
1277667
CC(C)(C)OC(=O)N1CCCC1C(=O)OC
C11H19NO4
MFCD00190828
boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate

Specifications

Yellow
73323-65-6
MFCD00190828
WVDGSSCWFMSRHN-MRVPVSSYSA-N
1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate
1277667
Clear Liquid at 20°C
N-(tert-Butoxycarbonyl)-D-proline Methyl Ester
1g
C11H19NO4
boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
CC(C)(C)OC(=O)N1CCCC1C(=O)OC
229.276
229.28
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator