missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Sulfinyl-p-toluenesulfonamide, TCI America™

Manufacturer:  TCI America T14615G

 View more versions of this product

Catalog No. T14615G



Specifications

Specifications

N-Sulfinyl-p-toluenesulfonamide
5g
C7H7NO3S2
N-Thionyl-p-toluenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)N=S=O
217.257
217.26
≥70.0% (T)
Undesignated
4104-47-6
MFCD00082512
VKTSIMMJOIPMGE-UHFFFAOYSA-N
4-methyl-N-sulfinylbenzenesulfonamide
562840
Crystal-Powder at 20°C

Chemical Identifiers

4104-47-6
217.257
VKTSIMMJOIPMGE-UHFFFAOYSA-N
562840
CC1=CC=C(C=C1)S(=O)(=O)N=S=O
C7H7NO3S2
MFCD00082512
N-Thionyl-p-toluenesulfonamide
4-methyl-N-sulfinylbenzenesulfonamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents