Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Succinyl-Gly-Gly-Phe-p-nitroanilide

Chemical Identifiers

Chemical Name or Material N-Succinyl-Gly-Gly-Phe-p-nitroanilide
CAS 68982-90-1
Molecular Formula C23H25N5O8
Formula Weight 499.47
MDL Number MFCD00038721
Synonym Suc-Gly-Gly-Phe-pNA; Chymotrypsin Substrate I, Colorimetric
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65375LB0

  SDS
alfa aesar™
J65375LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 499.47
Chemical Name or Material N-Succinyl-Gly-Gly-Phe-p-nitroanilide
CAS 68982-90-1
Synonym Suc-Gly-Gly-Phe-pNA; Chymotrypsin Substrate I, Colorimetric
Physical Form Solid
MDL Number MFCD00038721
Recommended Storage −30° to −10°C
Sensitivity Light Sensitive
Quantity 5mg
Color White to Off-white
Molecular Formula C23H25N5O8

  • Substrate for assay of chymotrypsin activity and for S. griseus protease B

Safety and Handling

−30° to −10°C