Learn More
N-Succinimidyl N-methylcarbamate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6356614
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
N-Succinimidyl N-methylcarbamate | |
C6H8N2O4 | |
25 g | |
Moisture sensitive | |
Slightly soluble in water. | |
CNC(=O)ON1C(=O)CCC1=O | |
172.14 | |
CHEBI:83949 | |
97% |
18342-66-0 | |
MFCD00800292 | |
1531863 | |
2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester | |
XMNGSPOWUCNRMO-UHFFFAOYSA-N | |
(2,5-dioxopyrrolidin-1-yl) N-methylcarbamate | |
4381935 | |
172.14 | |
Solid |
Chemical Identifiers
18342-66-0 | |
172.14 | |
XMNGSPOWUCNRMO-UHFFFAOYSA-N | |
4381935 | |
(2,5-dioxopyrrolidin-1-yl) N-methylcarbamate |
C6H8N2O4 | |
MFCD00800292 | |
2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester | |
CHEBI:83949 | |
CNC(=O)ON1C(=O)CCC1=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only