missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Succinimidyl N-methylcarbamate, 97%

Manufacturer:  Alfa Aesar™ H6356603

 View more versions of this product

Catalog No. AAH6356603



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Moisture sensitive
18342-66-0
MFCD00800292
1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-dioxopyrrolidin-1-yl methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester, mic substitute, n-succinimdyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimidyl-n-methylcarbamate
XMNGSPOWUCNRMO-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
4381935
172.14
97%
1g
C6H8N2O4
1531863
Slightly soluble in water.
CNC(=O)ON1C(=O)CCC1=O
172.14
CHEBI:83949
Solid

Chemical Identifiers

18342-66-0
172.14
XMNGSPOWUCNRMO-UHFFFAOYSA-N
4381935
(2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
C6H8N2O4
MFCD00800292
1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-dioxopyrrolidin-1-yl methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester, mic substitute, n-succinimdyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimidyl-n-methylcarbamate
CHEBI:83949
CNC(=O)ON1C(=O)CCC1=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Argon

Documents