missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl Ferrocenecarboxylate 98.0+%, TCI America™

Manufacturer:  TCI America S08201G

 View more versions of this product

Catalog No. S08201G



Specifications

Specifications

N-Succinimidyl Ferrocenecarboxylate
1g
C15H15FeNO4
Ferrocenecarboxylic Acid N-Succinimidyl Ester
C1CC(=O)N(C1=O)OC(=O)C2=CC=CC2.C1C=CC=C1.[Fe]
329.133
327.12
≥98.0% (HPLC)
Undesignated
115223-09-1
MFCD03427488
CNVNCOWBDQLYOU-UHFFFAOYSA-N
cyclopenta-1,3-diene;(2,5-dioxopyrrolidin-1-yl) cyclopenta-1,3-diene-1-carboxylate;iron
14226536
Crystal-Powder at 20°C

Chemical Identifiers

115223-09-1
329.133
CNVNCOWBDQLYOU-UHFFFAOYSA-N
14226536
C1CC(=O)N(C1=O)OC(=O)C2=CC=CC2.C1C=CC=C1.[Fe]
C15H15FeNO4
MFCD03427488
Ferrocenecarboxylic Acid N-Succinimidyl Ester
cyclopenta-1,3-diene;(2,5-dioxopyrrolidin-1-yl) cyclopenta-1,3-diene-1-carboxylate;iron
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents