missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl Acrylate 98.0+%, TCI America™

Manufacturer:  TCI America S08145G

 View more versions of this product

Catalog No. S08145G



Specifications

Specifications

N-Succinimidyl Acrylate
71°C
38862-24-7
MFCD00078261
YXMISKNUHHOXFT-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) prop-2-enoate
181508
Crystal-Powder at 20°C
White-Yellow
5g
C7H7NO4
1-acryloyloxy-2,5-pyrrolidinedione
C=CC(=O)ON1C(=O)CCC1=O
169.136
169.14
≥98.0% (GC)

Chemical Identifiers

38862-24-7
169.136
YXMISKNUHHOXFT-UHFFFAOYSA-N
181508
C=CC(=O)ON1C(=O)CCC1=O
C7H7NO4
MFCD00078261
1-acryloyloxy-2,5-pyrrolidinedione
(2,5-dioxopyrrolidin-1-yl) prop-2-enoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents