missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate 96.0+%, TCI America™

Manufacturer:  TCI America S08661G

 View more versions of this product

Catalog No. S08661G



Specifications

Specifications

N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate
1g
C14H9NO7
7-Hydroxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, N-Succinimidyl Umbelliferone-3-carboxylate, Umbelliferone-3-carboxylic Acid N-Succinimidyl Ester
C1CC(=O)N(C1=O)OC(=O)C2=CC3=C(C=C(C=C3)O)OC2=O
303.226
303.23
≥96.0% (HPLC,T)
Yellow
134471-24-2
MFCD00037568
KFEBWCYYRFZMTJ-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 7-hydroxy-2-oxochromene-3-carboxylate
5706554
Crystal-Powder at 20°C

Chemical Identifiers

134471-24-2
303.226
KFEBWCYYRFZMTJ-UHFFFAOYSA-N
5706554
C1CC(=O)N(C1=O)OC(=O)C2=CC3=C(C=C(C=C3)O)OC2=O
C14H9NO7
MFCD00037568
7-Hydroxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, N-Succinimidyl Umbelliferone-3-carboxylate, Umbelliferone-3-carboxylic Acid N-Succinimidyl Ester
(2,5-dioxopyrrolidin-1-yl) 7-hydroxy-2-oxochromene-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents