missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 6-Maleimidohexanoate 98.0+%, TCI America™

Manufacturer:  TCI America S0428100MG

 View more versions of this product

Catalog No. S0428100MG



Specifications

Specifications

N-Succinimidyl 6-Maleimidohexanoate [Cross-linking Reagent]
71°C
55750-63-5
MFCD00043043
VLARLSIGSPVYHX-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 6-(2,5-dioxopyrrol-1-yl)hexanoate
5091655
Crystal-Powder at 20°C
White-Yellow
100mg
C14H16N2O6
1h-pyrrole-2,5-dione, 1-6-2,5-dioxo-1-pyrrolidinyl oxy-6-oxohexyl
C1CC(=O)N(C1=O)OC(=O)CCCCCN2C(=O)C=CC2=O
308.29
308.29
≥98.0% (N)

Chemical Identifiers

55750-63-5
308.29
VLARLSIGSPVYHX-UHFFFAOYSA-N
5091655
C1CC(=O)N(C1=O)OC(=O)CCCCCN2C(=O)C=CC2=O
C14H16N2O6
MFCD00043043
1h-pyrrole-2,5-dione, 1-6-2,5-dioxo-1-pyrrolidinyl oxy-6-oxohexyl
(2,5-dioxopyrrolidin-1-yl) 6-(2,5-dioxopyrrol-1-yl)hexanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)

Documents