missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate, 98+%

Manufacturer:  Alfa Aesar™ H54627MB

 View more versions of this product

Catalog No. AAH54627MB



Specifications

Specifications

N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate
180°C to 182°C
64987-85-5
MFCD00009634
2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate
C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O
334.328
CHEBI:63174
≥98%
Light and moisture sensitive
25mg
C16H18N2O6
1555271
JJAHTWIKCUJRDK-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
125175
334.33

Chemical Identifiers

64987-85-5
334.328
JJAHTWIKCUJRDK-UHFFFAOYSA-N
125175
(2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
C16H18N2O6
MFCD00009634
2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate
CHEBI:63174
C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C; Store under Argon

Documents