missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 4-Maleimidobutyrate [Cross-linking Reagent], TCI America™

Manufacturer:  TCI America S03991G

 View more versions of this product

Catalog No. S03991G



Specifications

Specifications

N-Succinimidyl 4-Maleimidobutyrate [Cross-linking Reagent]
130°C
80307-12-6
MFCD00036817
PVGATNRYUYNBHO-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 4-(2,5-dioxopyrrol-1-yl)butanoate
133440
Crystal-Powder at 20°C
Undesignated
1g
C12H12N2O6
gmbs, 4-maleimidobutyric acid n-hydroxysuccinimide ester, 4-maleimidobutyric acid n-succinimidyl ester, n-gamma-maleimidobutyryloxy succinimide, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl butanoate, n-4-maleimidobutyryloxy succinimide, n-succinimidyl 4-maleimidobutyrate, n-succinimidyl-4-maleimidobutyrate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate
C1CC(=O)N(C1=O)OC(=O)CCCN2C(=O)C=CC2=O
280.236
280.24
≥98.0% (HPLC,N)

Chemical Identifiers

80307-12-6
280.236
PVGATNRYUYNBHO-UHFFFAOYSA-N
133440
C1CC(=O)N(C1=O)OC(=O)CCCN2C(=O)C=CC2=O
C12H12N2O6
MFCD00036817
gmbs, 4-maleimidobutyric acid n-hydroxysuccinimide ester, 4-maleimidobutyric acid n-succinimidyl ester, n-gamma-maleimidobutyryloxy succinimide, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl butanoate, n-4-maleimidobutyryloxy succinimide, n-succinimidyl 4-maleimidobutyrate, n-succinimidyl-4-maleimidobutyrate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate
(2,5-dioxopyrrolidin-1-yl) 4-(2,5-dioxopyrrol-1-yl)butanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)

Documents