missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 4-Benzoylbenzoate 98.0+%, TCI America™

Manufacturer:  TCI America S0863200MG

 View more versions of this product

Catalog No. S0863200MG



Specifications

Specifications

N-Succinimidyl 4-Benzoylbenzoate
209°C
91990-88-4
MFCD00058572
MVQNJLJLEGZFGP-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 4-benzoylbenzoate
2762595
Crystal-Powder at 20°C
White
200mg
C18H13NO5
4-Benzoylbenzoic Acid N-Succinimidyl Ester
C1CC(=O)N(C1=O)OC(=O)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3
323.304
323.30
≥98.0% (HPLC,N)

Chemical Identifiers

91990-88-4
323.304
MVQNJLJLEGZFGP-UHFFFAOYSA-N
2762595
C1CC(=O)N(C1=O)OC(=O)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3
C18H13NO5
MFCD00058572
4-Benzoylbenzoic Acid N-Succinimidyl Ester
(2,5-dioxopyrrolidin-1-yl) 4-benzoylbenzoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents