missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 3-Maleimidopropionate [Cross-linking Reagent], TCI America™

Manufacturer:  TCI America S04275G

 View more versions of this product

Catalog No. S04275G



Specifications

Specifications

N-Succinimidyl 3-Maleimidopropionate [Cross-linking Reagent]
168°C
55750-62-4
MFCD00043141
JKHVDAUOODACDU-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
4620597
Crystal-Powder at 20°C
White-Yellow
5g
C11H10N2O6
n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate
C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O
266.209
266.21
≥98.0% (HPLC,N)

Chemical Identifiers

55750-62-4
266.209
JKHVDAUOODACDU-UHFFFAOYSA-N
4620597
C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O
C11H10N2O6
MFCD00043141
n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents