missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 3-maleimidobenzoate, 95%, Thermo Scientific Chemicals

A heterobifunctional coupling reagent useful for forming enzyme immunoconjugates

Supplier:  Thermo Scientific Chemicals J61410MD

 View more versions of this product

Catalog No. AAJ61410MD



Description

Description

N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates

Solubility
Soluble in DMF or ethyl acetate.

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

N-Succinimidyl 3-maleimidobenzoate
175°C to 177°C
C15H10N2O6
250 mg
Light and moisture sensitive
Soluble in DMF or ethyl acetate.
C1CC(=O)N(C1=O)OC(=O)C2=CC(=CC=C2)N3C(=O)C=CC3=O
314.253
314.26
Powder
58626-38-3
White
MFCD00005514
1505254
3-maleimidobenzoic acid n-hydroxysuccinimide ester, n-succinimidyl 3-maleimidobenzoate, 3-maleimidobenzoyl n-hydroxysuccinimide, 1-3-2,5-dioxopyrrolidinyl oxy carbonyl phenyl-1h-pyrrole-2,5-dione, m-maleimidobenzoyl-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl benzoate, 3-n-maleimidobenzoic acid n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl benzoate, ambotzpeg2090
LLXVXPPXELIDGQ-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)benzoate
93861
95%

Chemical Identifiers

58626-38-3
314.253
LLXVXPPXELIDGQ-UHFFFAOYSA-N
93861
C1CC(=O)N(C1=O)OC(=O)C2=CC(=CC=C2)N3C(=O)C=CC3=O
C15H10N2O6
MFCD00005514
3-maleimidobenzoic acid n-hydroxysuccinimide ester, n-succinimidyl 3-maleimidobenzoate, 3-maleimidobenzoyl n-hydroxysuccinimide, 1-3-2,5-dioxopyrrolidinyl oxy carbonyl phenyl-1h-pyrrole-2,5-dione, m-maleimidobenzoyl-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl benzoate, 3-n-maleimidobenzoic acid n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl benzoate, ambotzpeg2090
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)benzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 261-368-8

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only