Please login to your online account to display your discounted pricing

N-Stearoyl-D-erythro-sphingosine, 99+%, synthetical, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Stearoyl-D-erythro-sphingosine
Name Note 99+%
CAS 2304-81-6
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C36H71NO3
Formula Weight 565.96
MDL Number MFCD02259139
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC331550500

  SDS
thermo fisher scientific
331550500
Glass bottle 50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Powder
Formula Weight 565.96
Quantity 50mg
Color White
Melting Point 99° to 101°C
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym Ceramide C18
Packaging Glass bottle
Chemical Name or Material N-Stearoyl-D-erythro-sphingosine
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C36H71NO3
MDL Number MFCD02259139
CAS 2304-81-6