Please login to your online account to display your discounted pricing

N-Stearoyl-D-erythro-sphingosine, 99+%, synthetical, ACROS Organics™


Quantity 50mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  331550500

 View more versions of this product

Catalog No. AC331550500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 99° to 101°C
Quantity 50mg
Chemical Name or Material N-Stearoyl-D-erythro-sphingosine
Name Note 99+%
CAS 2304-81-6
CAS Min % 99.0
CAS Max % 100.0
Physical Form White powder
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C36H71NO3
Formula Weight 565.96g/mol
MDL Number MFCD02259139
Synonym Ceramide C18

Specifications

Chemical Name or Material N-Stearoyl-D-erythro-sphingosine
Name Note 99+%
CAS 2304-81-6
CAS Min % 99.0
CAS Max % 100.0
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C36H71NO3
Formula Weight 565.96g/mol
MDL Number MFCD02259139
Synonym Ceramide C18