Please login to your online account to display your discounted pricing

Alfa Aesar™ N-[(S)-alpha-(4-Chlorophenyl)benzyl]formamide, 98%

Chemical Identifiers

Chemical Name or Material N-[(S)-alpha-(4-Chlorophenyl)benzyl]formamide
CAS 474654-19-8
Formula Weight 245.7
Synonym N-[(S)-(4-Chlorophenyl)phenylmethyl]formamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6141303

 
alfa aesar™
H6141303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61413MD

 
alfa aesar™
H61413MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.7
Synonym N-[(S)-(4-Chlorophenyl)phenylmethyl]formamide
Assay 98%
CAS 474654-19-8
Physical Form Solid
Quantity 1g
Chemical Name or Material N-[(S)-alpha-(4-Chlorophenyl)benzyl]formamide