Please login to your online account to display your discounted pricing

N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  332250050

 View more versions of this product

Catalog No. AC332250050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 279.34
Quantity 5g
Color White
Melting Point 148° to 153°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
Packaging Glass bottle
Name Note 97%
Molecular Formula C15H21NO4
MDL Number MFCD00209976
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 97%
CAS 82717-96-2

Specifications

Chemical Name or Material N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
Name Note 97%
CAS 82717-96-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C15H21NO4
Formula Weight 279.34
MDL Number MFCD00209976
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents