Please login to your online account to display your discounted pricing

N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
Name Note 97%
CAS 82717-96-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C15H21NO4
Formula Weight 279.34
MDL Number MFCD00209976
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC332250010

  SDS
thermo fisher scientific
332250010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC332250050

  SDS
thermo fisher scientific
332250050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 279.34
Physical Form Crystalline Powder
Quantity 1g
Color White
Melting Point 148° to 153°C
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
CAS 82717-96-2
Molecular Formula C15H21NO4
Name Note 97%
Chemical Name or Material N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
MDL Number MFCD00209976
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.