Please login to your online account to display your discounted pricing

Alfa Aesar™ N-[(R)-alpha-(4-Chlorophenyl)benzyl]formamide, 98%

Chemical Identifiers

Chemical Name or Material N-[(R)-alpha-(4-Chlorophenyl)benzyl]formamide
CAS 474654-18-7
Formula Weight 245.7
Synonym N-[(R)-(4-Chlorophenyl)phenylmethyl]formamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH61607MD

 
alfa aesar™
H61607MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6160703

 
alfa aesar™
H6160703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.7
Chemical Name or Material N-[(R)-alpha-(4-Chlorophenyl)benzyl]formamide
Physical Form Solid
Assay 98%
CAS 474654-18-7
Synonym N-[(R)-(4-Chlorophenyl)phenylmethyl]formamide
Quantity 250mg