missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propylurea, 98%

Manufacturer:  Alfa Aesar™ A1016006

 View more versions of this product

Catalog No. AAA1016006



Description

Description

Applications
n-Propylurea is used as a reactant in the preparation of N-1 monosubstituted 8-pyrazolyl xanthines as high affinity A2B adenosine receptor antagonists.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

n-Propylurea
5g
C4H10N2O
MFCD00014793
ZQZJKHIIQFPZCS-UHFFFAOYSA-N
propylurea
12303
98%
99°C to 104°C
627-06-5
CH3CH2CH2NHCONH2
n-propylurea, 1-propylurea, urea, propyl, urea, n-propyl, propyl urea, unii-0pk51j4av7, n-propyl urea, n'-propylurea, amino-n-propylamide, acmc-1ato6
CCCNC(=O)N
102.137
102.14

Chemical Identifiers

627-06-5
102.137
ZQZJKHIIQFPZCS-UHFFFAOYSA-N
12303
CCCNC(=O)N
C4H10N2O
MFCD00014793
n-propylurea, 1-propylurea, urea, propyl, urea, n-propyl, propyl urea, unii-0pk51j4av7, n-propyl urea, n'-propylurea, amino-n-propylamide, acmc-1ato6
propylurea
Safety and Handling

Safety and Handling

RTECSNumber : YU151550

TSCA : TSCA

SDS Documents