missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propylurea, 98%

$42.72 - $468.81

Chemical Identifiers

CAS 627-06-5
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00014793
InChI Key ZQZJKHIIQFPZCS-UHFFFAOYSA-N
Synonym n-propylurea, 1-propylurea, urea, propyl, urea, n-propyl, propyl urea, unii-0pk51j4av7, n-propyl urea, n'-propylurea, amino-n-propylamide, acmc-1ato6
PubChem CID 12303
IUPAC Name propylurea
SMILES CCCNC(=O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1016006 View Documents Alfa Aesar™
A1016006
5g Each for $42.72
 
AAA1016022 View Documents Alfa Aesar™
A1016022
100g Each for $468.81
 
AAA1016014 View Documents Alfa Aesar™
A1016014
25g Each for $145.88
 
Description

Description

Applications
n-Propylurea is used as a reactant in the preparation of N-1 monosubstituted 8-pyrazolyl xanthines as high affinity A2B adenosine receptor antagonists.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

627-06-5
102.137
ZQZJKHIIQFPZCS-UHFFFAOYSA-N
12303
CCCNC(=O)N
C4H10N2O
MFCD00014793
n-propylurea, 1-propylurea, urea, propyl, urea, n-propyl, propyl urea, unii-0pk51j4av7, n-propyl urea, n'-propylurea, amino-n-propylamide, acmc-1ato6
propylurea

Specifications

99°C to 104°C
627-06-5
CH3CH2CH2NHCONH2
n-propylurea, 1-propylurea, urea, propyl, urea, n-propyl, propyl urea, unii-0pk51j4av7, n-propyl urea, n'-propylurea, amino-n-propylamide, acmc-1ato6
CCCNC(=O)N
102.137
102.14
n-Propylurea
5g
C4H10N2O
MFCD00014793
ZQZJKHIIQFPZCS-UHFFFAOYSA-N
propylurea
12303
98%
Safety and Handling

Safety and Handling

RTECSNumber : YU151550

TSCA : TSCA