missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propylurea, 98%

Manufacturer:  Alfa Aesar™ A1016014

 View more versions of this product

Catalog No. AAA1016014



Description

Description

Applications
n-Propylurea is used as a reactant in the preparation of N-1 monosubstituted 8-pyrazolyl xanthines as high affinity A2B adenosine receptor antagonists.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

n-Propylurea
25g
98%
CH3CH2CH2NHCONH2
ZQZJKHIIQFPZCS-UHFFFAOYSA-N
propylurea
12303
99°C to 104°C
627-06-5
C4H10N2O
MFCD00014793
CCCNC(=O)N
102.137
102.14

Chemical Identifiers

627-06-5
102.137
ZQZJKHIIQFPZCS-UHFFFAOYSA-N
propylurea
C4H10N2O
MFCD00014793
12303
CCCNC(=O)N
Safety and Handling

Safety and Handling

RTECSNumber : YU151550

TSCA : TSCA

SDS Documents