missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Propyltriphenylphosphonium bromide, 99%, Acros Organics™

Manufacturer:  Acros Organics 157631000

 View more versions of this product

Catalog No. AC157631000


 

Description

Description

For C-C Bond Formation (Olefination)
Specifications

Specifications

n-Propyltriphenylphosphonium bromide
99%
98.5
Authentic
99%
C21H23BrP+
MFCD00011843
XMQSELBBYSAURN-UHFFFAOYSA-N
CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.Br
386.293
385.27
100g
15912-75-1
100.0
Plastic bottle
99%
CH3(CH2)2P(C6H5)3Br
White
235.0°C to 239.0°C
triphenyl(propyl)phosphanium;hydrobromide
88526383
Crystalline Powder

Chemical Identifiers

15912-75-1
386.293
XMQSELBBYSAURN-UHFFFAOYSA-N
C21H23BrP+
MFCD00011843
88526383
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 240-056-5

TSCA : TSCA

SDS Documents