missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Propyltriphenylphosphonium bromide, 99%, Acros Organics™

Manufacturer:  Thermo Fisher Scientific 157630250

 View more versions of this product

Catalog No. AC157630250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

For C-C Bond Formation (Olefination)
Specifications

Specifications

n-Propyltriphenylphosphonium bromide
Plastic bottle
235.0°C to 239.0°C
99%
98.5
98.5% min. (Argentometry)
CH3(CH2)2P(C6H5)3Br
propyl triphenyl phosphonium bromide
XMQSELBBYSAURN-UHFFFAOYSA-N
triphenyl(propyl)phosphanium;hydrobromide
88526383
Crystalline Powder
Authentic
White
25g
15912-75-1
100.0
C21H22BrP
MFCD00011843
Solubility in water: soluble. Other solubilities: soluble in chloroform and polar solvents
CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.Br
385.27
385.27
99%

Chemical Identifiers

15912-75-1
385.27
XMQSELBBYSAURN-UHFFFAOYSA-N
88526383
CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.Br
C21H22BrP
MFCD00011843
propyl triphenyl phosphonium bromide
triphenyl(propyl)phosphanium;hydrobromide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 240-056-5

TSCA : TSCA

SDS Documents