Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propyl methacrylate, 95%, stab. with 200ppm 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material n-Propyl methacrylate
CAS 2210-28-8
Name Note stabilized with 200ppm 4-methoxyphenol
Molecular Formula C7H12O2
Formula Weight 128.17
MDL Number MFCD00048690
UN Number UN3272
Beilstein 1744849
Synonym Methacrylic acid n-propyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0949306

  SDS
alfa aesar™
L0949306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0949314

  SDS
alfa aesar™
L0949314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.17
Chemical Name or Material n-Propyl methacrylate
Molecular Formula C7H12O2
MDL Number MFCD00048690
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note stabilized with 200ppm 4-methoxyphenol
RTECS Number OZ5250000
Synonym Methacrylic acid n-propyl ester
CAS 2210-28-8
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Assay 95%
Health Hazard 3 GHS P Statement
P305+P351+P338-P302+P352
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Odor Pungent
Flash Point 30°C (86°F)
UN Number UN3272
Quantity 5g
Boiling Point 140° to 141°C
Beilstein 1744849
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P305+P351+P338-P302+P352
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.