Please login to your online account to display your discounted pricing

Nω-Propyl-L-arginine hydrochloride, Tocris


Highly selective inhibitor of nNOS

Quantity 10mg

Manufacturer: r & d systems  RDS120010

 View more versions of this product

Catalog No. 120010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 252.74
Quantity 10mg
Molecular Formula C9H20N4O2·HCl
Assay Percent Range >98%
Synonym N5-[Imino(propylamino)methyl]-L-ornithine hydrochloride
CAS 137361-05-8
Chemical Name or Material Nω-Propyl-L-arginine hydrochloride
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material Nω-Propyl-L-arginine hydrochloride
Assay Percent Range >98%
Molecular Formula C9H20N4O2·HCl
Formula Weight 252.74
Synonym N5-[Imino(propylamino)methyl]-L-ornithine hydrochloride
Accessories

Desiccate at Room Temperature