missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nω-Propyl-L-arginine hydrochloride, Tocris

Highly selective inhibitor of nNOS

$225.77 - $924.23

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C9H20N4O2·HCl
Formula Weight 252.74
Synonym N5-[Imino(propylamino)methyl]-L-ornithine hydrochloride
Chemical Name or Material Nω-Propyl-L-arginine hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
120050 R & D Systems
RDS120050
50mg Each for $924.23
120010 R & D Systems
RDS120010
10mg Each for $225.77
Specifications Description & Specifications

Chemical Identifiers

>98%
252.74
Nω-Propyl-L-arginine hydrochloride
C9H20N4O2·HCl
N5-[Imino(propylamino)methyl]-L-ornithine hydrochloride

Specifications

137361-05-8
C9H20N4O2·HCl
N5-[Imino(propylamino)methyl]-L-ornithine hydrochloride
Nω-Propyl-L-arginine hydrochloride
>98%
252.74
50mg

Safety and Handling

Recommended Storage : Desiccate at Room Temperature