missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Propyl isocyanate, 99%, ACROS Organics™

Chemical Identifiers

CAS 110-78-1
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00002045
InChI Key OQURWGJAWSLGQG-UHFFFAOYSA-N
Synonym 1-isocyanato-propane, 1-propyl isocyanate, 1-propylisocyanate, isocyanic acid propyl ester, isocyanic acid, propyl ester, m-propyl isocyanate, n-propyl isocyanate, propane, 1-isocyanato, propyl isocyanate, propylisocyanate
PubChem CID 61033
IUPAC Name 1-isocyanatopropane
SMILES CCCN=C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC131600250
SDS View Documents
Thermo Fisher Scientific
AC131600250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC131601000
SDS View Documents
Thermo Fisher Scientific
AC131601000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC131605000
SDS View Documents
Thermo Fisher Scientific
AC131605000
500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

110-78-1
85.11
OQURWGJAWSLGQG-UHFFFAOYSA-N
61033
CCCN=C=O
C4H7NO
MFCD00002045
1-isocyanato-propane, 1-propyl isocyanate, 1-propylisocyanate, isocyanic acid propyl ester, isocyanic acid, propyl ester, m-propyl isocyanate, n-propyl isocyanate, propane, 1-isocyanato, propyl isocyanate, propylisocyanate
1-isocyanatopropane

Specifications

0.8948g/mL
Authentic
1.3940 to 1.3970
83.0°C to 84.0°C
-30.0°C
110-78-1
100.0
C4H7NO
MFCD00002045
1-isocyanato-propane, 1-propyl isocyanate, 1-propylisocyanate, isocyanic acid propyl ester, isocyanic acid, propyl ester, m-propyl isocyanate, n-propyl isocyanate, propane, 1-isocyanato, propyl isocyanate, propylisocyanate
OQURWGJAWSLGQG-UHFFFAOYSA-N
1-isocyanatopropane
61033
Liquid
n-Propyl isocyanate, 99%
−1°C
Glass bottle
0.8948
Yellow
25g
98.5
98.5% min. (GC)
CH3CH2CH2NCO
04,I,366
Solubility in water: reacts vigorously
CCCN=C=O
85.11
85.11
99%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-803-6

Recommended Storage : Product may develop some turbidity or precipitate

SDS