Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propyl hexanoate, 98%

Chemical Identifiers

Chemical Name or Material n-Propyl hexanoate
CAS 626-77-7
Molecular Formula C9H18O2
Linear Formula CH3(CH2)4CO2CH2CH2CH3
Formula Weight 158.24
MDL Number MFCD00053803
UN Number UN3272
Synonym n-Propyl caproate; Hexanoic acid n-propyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2521514

  SDS
alfa aesar™
B2521514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2521522

  SDS
alfa aesar™
B2521522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2521536

  SDS
alfa aesar™
B2521536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.24
Chemical Name or Material n-Propyl hexanoate
TSCA TSCA
Assay 98%
Molecular Formula C9H18O2
Linear Formula CH3(CH2)4CO2CH2CH2CH3
MDL Number MFCD00053803
CAS 626-77-7
UN Number UN3272
Synonym n-Propyl caproate; Hexanoic acid n-propyl ester
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Flash Point 52°C (126°F)
Odor Fruit-like
Boiling Point 187°C
Melting Point −69°C
Quantity 25g
Safety and Handling

Warning