Please login to your online account to display your discounted pricing

n-Propyl Ether, 99+%, ACROS Organics™

$297.52 - $297.52

Chemical Identifiers

Chemical Name or Material n-Propyl ether
Name Note 99+%
CAS 111-43-3
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H14O
Linear Formula (CH3CH2CH2)2O
Formula Weight 102.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC150040500

  SDS
thermo fisher scientific
150040500
Glass bottle 50mL Each for $297.52
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Liquid
Formula Weight 102.18
Quantity 50mL
Color Clear colorless
EINECS Number 203-869-6
TSCA TSCA
Density 0.7300g/mL
RTECS Number UJ5125000
Beilstein 01, 354
Merck Index 15, 3385
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material n-Propyl ether
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3790 to 1.3810 (20°C, 589nm)
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C6H14O
Linear Formula (CH3CH2CH2)2O
MDL Number MFCD00009376
CAS 111-43-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.