Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propyl cinnamate, 98%

$48.49 - $172.41

Chemical Identifiers

Chemical Name or Material n-Propyl cinnamate
CAS 7778-83-8
Molecular Formula C6H5CH=CHCO2(CH2)2CH3
Formula Weight 190.24
MDL Number MFCD00051556
Beilstein 2044816
Synonym Cinnamic acid n-propyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0787118

 
alfa aesar™
L0787118
50g Each for $48.49

AAL0787130

 
alfa aesar™
L0787130
250g Each for $172.41
Description & Specifications

Specifications

Formula Weight 190.24
Chemical Name or Material n-Propyl cinnamate
Molecular Formula C6H5CH=CHCO2(CH2)2CH3
MDL Number MFCD00051556
CAS 7778-83-8
TSCA TSCA
Assay 98%
Beilstein 2044816
Synonym Cinnamic acid n-propyl ester
RTECS Number GE0525000
Flash Point >99°C (210°F)
Quantity 50g
Boiling Point 282° to 284°C