Please login to your online account to display your discounted pricing

n-Propyl Alcohol AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material n-Propyl Alcohol
CAS 71-23-8
Molecular Formula CH3(CH2)2OH
Formula Weight 60.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7169500

 
avantor performance
716904
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK716920

 
avantor performance
716919
Steel Pail 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK71694

 
avantor performance
716908
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.1
Quantity 500mL
Chemical Name or Material n-Propyl Alcohol
Molecular Formula CH3(CH2)2OH
CAS 71-23-8
Packaging Glass