Please login to your online account to display your discounted pricing

n-Propyl Alcohol AR, Macron Fine Chemicals™


macron

Quantity 4L
Packaging Glass

Manufacturer: avantor performance  716908

 View more versions of this product

Catalog No. MK71694

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 60.1
Quantity 4L
Molecular Formula CH3(CH2)2OH
CAS 71-23-8
Chemical Name or Material n-Propyl Alcohol
Packaging Glass

Specifications

Chemical Name or Material n-Propyl Alcohol
CAS 71-23-8
Molecular Formula CH3(CH2)2OH
Formula Weight 60.1