Learn More
n-Propyl acrylate, 95%, stab. with 0.1% 4-methoxyphenol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0971418
Description
n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
n-Propyl acrylate | |
925-60-0 | |
43°C to 44°C (40 mmHg) | |
C6H10O2 | |
MFCD00048693 | |
UN3272 | |
n-propyl acrylate, propyl acrylate, 2-propenoic acid, propyl ester, propyl 2-propenoate, acrylic acid, propyl ester, propylacrylate, ccris 8584, 1-propyl acrylate, acrylic acid propyl, acrylic acid propyl ester | |
PNXMTCDJUBJHQJ-UHFFFAOYSA-N | |
propyl prop-2-enoate | |
13550 | |
95% |
stabilized with 0.1% 4-methoxyphenol | |
0.92 | |
24°C (75°F) | |
1.4123 | |
50 g | |
1744954 | |
Immiscible with water. | |
CCCOC(=O)C=C | |
114.14 | |
114.15 |
Chemical Identifiers
925-60-0 | |
114.14 | |
PNXMTCDJUBJHQJ-UHFFFAOYSA-N | |
13550 | |
CCCOC(=O)C=C |
C6H10O2 | |
MFCD00048693 | |
n-propyl acrylate, propyl acrylate, 2-propenoic acid, propyl ester, propyl 2-propenoate, acrylic acid, propyl ester, propylacrylate, ccris 8584, 1-propyl acrylate, acrylic acid propyl, acrylic acid propyl ester | |
propyl prop-2-enoate |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 213-120-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only