missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propyl 4-hydroxybenzoate, 99+%

$56.74 - $536.66

Chemical Identifiers

CAS 94-13-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002354
InChI Key QELSKZZBTMNZEB-UHFFFAOYSA-N
Synonym propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept
PubChem CID 7175
ChEBI CHEBI:32063
IUPAC Name propyl 4-hydroxybenzoate
SMILES CCCOC(=O)C1=CC=C(C=C1)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA101800I View Documents Alfa Aesar™
A101800I
5000g Each for $536.66
 
AAA101800B View Documents Alfa Aesar™
A101800B
1000g Each for $137.24
 
AAA1018030 View Documents Alfa Aesar™
A1018030
250g Each for $56.74
 
Description

Description

Applications
n-Propyl 4-hydroxybenzoate is used as antimicrobial preservatives in pharmaceuticals, foods, cosmetics and shampoos. It is widely used as preservatives in personal care products like creams, lotions and bath products. Further, it is used as a food additive. Its derivative, sodium propyl p-hydroxybenzoate is used as an anti-fungal preservation agent.

Solubility
Soluble in propylene glycol, ethanol, ether, acetone, methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and alkali metals.
Specifications

Chemical Identifiers

94-13-3
180.203
QELSKZZBTMNZEB-UHFFFAOYSA-N
7175
propyl 4-hydroxybenzoate
C10H12O3
MFCD00002354
propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept
CHEBI:32063
CCCOC(=O)C1=CC=C(C=C1)O

Specifications

180°C (356°F)
5000g
C10H12O3
1103245
propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept
CCCOC(=O)C1=CC=C(C=C1)O
180.203
CHEBI:32063
≥99%
95°C to 99°C
94-13-3
MFCD00002354
14,7866
QELSKZZBTMNZEB-UHFFFAOYSA-N
propyl 4-hydroxybenzoate
7175
180.2
n-Propyl 4-hydroxybenzoate
Safety and Handling

Safety and Handling

RTECSNumber : DH2800000

TSCA : TSCA