missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Propyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0906618

 View more versions of this product

Catalog No. AAL0906618



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Propyl 4-aminobenzoate
73°C to 75°C
MFCD00017111
1211203
NBFQYHKHPBMJJV-UHFFFAOYSA-N
propyl 4-aminobenzoate
7174
98%
94-12-2
C10H13NO2
50 g
risocaine, propaesin, propazyl, propesine, propylcain, raythesin, keloform p, propyl p-aminobenzoate, propesin, 4-propoxycarbonyl aniline
CCCOC(=O)C1=CC=C(N)C=C1
179.22
179.22

Chemical Identifiers

94-12-2
179.22
NBFQYHKHPBMJJV-UHFFFAOYSA-N
7174
CCCOC(=O)C1=CC=C(N)C=C1
C10H13NO2
MFCD00017111
risocaine, propaesin, propazyl, propesine, propylcain, raythesin, keloform p, propyl p-aminobenzoate, propesin, 4-propoxycarbonyl aniline
propyl 4-aminobenzoate
Safety and Handling

Safety and Handling

EINECSNumber : 202-306-1

RTECSNumber : DG3090000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only