Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propyl 2-methylvalerate, 98%

Chemical Identifiers

Chemical Name or Material n-Propyl 2-methylvalerate
CAS 6639-14-1
Molecular Formula C9H18O2
Formula Weight 158.24
MDL Number MFCD01632135
UN Number UN1993
Synonym 2-Methylvaleric acid n-propyl ester; n-Propyl 2-methylpentanoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2132514

 
alfa aesar™
B2132514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2132522

 
alfa aesar™
B2132522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.24
Chemical Name or Material n-Propyl 2-methylvalerate
Assay 98%
Molecular Formula C9H18O2
MDL Number MFCD01632135
CAS 6639-14-1
UN Number UN1993
Synonym 2-Methylvaleric acid n-propyl ester; n-Propyl 2-methylpentanoate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 51°C (124°F)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.