Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Propyl 2-ethylhexanoate, 99%

$44.00 - $171.77

Chemical Identifiers

Chemical Name or Material n-Propyl 2-ethylhexanoate
CAS 172354-89-1
Molecular Formula CH3(CH2)3CH(C2H5)CO2CH2CH2CH3
Formula Weight 186.29
MDL Number MFCD00048820
Synonym 2-Ethylhexanoic acid n-propyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2041922

  SDS
alfa aesar™
B2041922
100g Each for $44.00

AAB2041936

  SDS
alfa aesar™
B2041936
500g Each for $171.77
Description & Specifications

Specifications

Formula Weight 186.29
Quantity 100g
MDL Number MFCD00048820
Synonym 2-Ethylhexanoic acid n-propyl ester
Molecular Formula CH3(CH2)3CH(C2H5)CO2CH2CH2CH3
Chemical Name or Material n-Propyl 2-ethylhexanoate
CAS 172354-89-1
Assay 99%