missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Propargylphthalimide, TCI America™

Manufacturer:  TCI America P23295G

 View more versions of this product

Catalog No. P23295G



Specifications

Specifications

N-Propargylphthalimide
153°C
7223-50-9
C11H7NO2
N-(2-Propynyl)phthalimide
C#CCN1C(=O)C2=CC=CC=C2C1=O
185.182
185.18
Undesignated
5g
≥98.0% (GC)
MFCD00065028
PAZCLCHJOWLTGA-UHFFFAOYSA-N
2-prop-2-ynylisoindole-1,3-dione
81644
Crystal-Powder at 20°C

Chemical Identifiers

7223-50-9
185.182
PAZCLCHJOWLTGA-UHFFFAOYSA-N
81644
C#CCN1C(=O)C2=CC=CC=C2C1=O
C11H7NO2
MFCD00065028
N-(2-Propynyl)phthalimide
2-prop-2-ynylisoindole-1,3-dione
Safety and Handling

Safety and Handling

TSCA : No

Documents