missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Propargylphthalimide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5342906

 View more versions of this product

Catalog No. AAH5342906



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Propargylphthalimide
148°C to 152°C
MFCD00065028
n-propargylphthalimide, 2-prop-2-yn-1-yl isoindoline-1,3-dione, n-prop-2-ynylphthalimide, 2-2-propynyl-1h-isoindole-1,3 2h-dione, 2-prop-2-ynyl-isoindole-1,3-dione, n-propargyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-2-propynyl, 2-prop-2-ynyl isoindoline-1,3-dione, 2-prop-2-yn-1-yl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-2-propyn-1-yl
O=C1N(CC#C)C(=O)C2=CC=CC=C12
185.18
185.18
7223-50-9
C11H7NO2
5 g
PAZCLCHJOWLTGA-UHFFFAOYSA-N
2-prop-2-ynylisoindole-1,3-dione
81644
98%

Chemical Identifiers

7223-50-9
185.18
PAZCLCHJOWLTGA-UHFFFAOYSA-N
81644
O=C1N(CC#C)C(=O)C2=CC=CC=C12
C11H7NO2
MFCD00065028
n-propargylphthalimide, 2-prop-2-yn-1-yl isoindoline-1,3-dione, n-prop-2-ynylphthalimide, 2-2-propynyl-1h-isoindole-1,3 2h-dione, 2-prop-2-ynyl-isoindole-1,3-dione, n-propargyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-2-propynyl, 2-prop-2-ynyl isoindoline-1,3-dione, 2-prop-2-yn-1-yl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-2-propyn-1-yl
2-prop-2-ynylisoindole-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H500

EINECSNumber : 230-621-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only