Please login to your online account to display your discounted pricing

N-Propargylphthalimide 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Propargylphthalimide
CAS 7223-50-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H7NO2
Formula Weight 185.18
MDL Number MFCD00065028
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381350010

  SDS
fisher scientific acros
AC381350010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 185.18
Physical Form Powder
Quantity 1g
Color Beige to orange
Melting Point 145° to 153°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material N-Propargylphthalimide
Assay Percent Range 97%
Packaging Glass bottle
MDL Number MFCD00065028
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Molecular Formula C11H7NO2
CAS 7223-50-9
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.