Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Propargyloxyphthalimide, 97%

$98.39 - $98.39

Chemical Identifiers

Chemical Name or Material N-Propargyloxyphthalimide
CAS 4616-63-1
Molecular Formula C11H7NO3
Formula Weight 201.18
MDL Number MFCD00005890
Beilstein 1530730
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0923306

  SDS
alfa aesar™
L0923306
5g Each for $98.39
Description & Specifications

Specifications

Formula Weight 201.18
Chemical Name or Material N-Propargyloxyphthalimide
CAS 4616-63-1
MDL Number MFCD00005890
Molecular Formula C11H7NO3
Assay 97%
Beilstein 1530730
Melting Point 150° to 152°C
Quantity 5g