Please login to your online account to display your discounted pricing

N-(Propargyloxy)phthalimide 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  131490250

 View more versions of this product

Catalog No. AC131490250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 201.18
Quantity 25g
Color Beige-brown
EINECS Number 225-024-0
Melting Point 149° to 152°C
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 97.5
Packaging Glass bottle
Molecular Formula C11H7NO3
Name Note 98%
Chemical Name or Material N-(Propargyloxy)phthalimide
MDL Number MFCD00005890
CAS 4616-63-1
Assay Percent Range 98%

Specifications

Chemical Name or Material N-(Propargyloxy)phthalimide
Name Note 98%
CAS 4616-63-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H7NO3
Formula Weight 201.18
MDL Number MFCD00005890