Please login to your online account to display your discounted pricing

N-Prop-2-ynylurea, Maybridge

Chemical Identifiers

Chemical Name or Material N-Prop-2-ynylurea
CAS 5221-62-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H6N2O
Linear Formula H2NC(O)NHCH2Cidentical toCH
Formula Weight 98.1
MDL Number MFCD00052817
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CD11100DE

 
fisher scientific company
CD11100DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CD11100EA

 
fisher scientific company
CD11100EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.1
Quantity 5g
Assay 97%
CAS Max % 100.0
Chemical Name or Material N-Prop-2-ynylurea
Assay Percent Range 97%
Linear Formula H2NC(O)NHCH2Cidentical toCH
MDL Number MFCD00052817
CAS 5221-62-5
CAS Min % 96.5
Molecular Formula C4H6N2O