Please login to your online account to display your discounted pricing

N-Prop-2-ynylurea, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CD11100EA

 View more versions of this product

Catalog No. CD11100EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 98.1
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material N-Prop-2-ynylurea
Linear Formula H2NC(O)NHCH2Cidentical toCH
CAS 5221-62-5
Molecular Formula C4H6N2O
CAS Min % 96.5
MDL Number MFCD00052817
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material N-Prop-2-ynylurea
CAS 5221-62-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H6N2O
Linear Formula H2NC(O)NHCH2Cidentical toCH
Formula Weight 98.1
MDL Number MFCD00052817