Please login to your online account to display your discounted pricing

N-Pivaloyl-o-benzylaniline, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material N-Pivaloyl-o-benzylaniline
Name Note 99%
CAS 85864-33-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C18H21NO
Formula Weight 267.37
MDL Number MFCD00075427
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC346740010

  SDS
thermo fisher scientific
346740010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 267.37
Physical Form Powder
Quantity 1g
Color White
Melting Point 86° to 90°C
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym 2'-Benzyl-2, 2-dimethylpropionanilide
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Molecular Formula C18H21NO
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Chemical Name or Material N-Pivaloyl-o-benzylaniline
Name Note 99%
MDL Number MFCD00075427
CAS 85864-33-1
Assay Percent Range 99%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.