missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Phthalyl-L-tryptophan 98.0+%, TCI America™
Supplier: TCI America P202350MG
Specifications
N-Phthalyl-L-tryptophan | |
181°C | |
C19H14N2O4 | |
50 mg | |
HPTXLHAHLXOAKV-MRXNPFEDSA-N | |
(2R)-2-(1,3-dioxoisoindol-2-yl)-3-(1H-indol-3-yl)propanoic acid | |
702560 | |
≥98.0% (HPLC,T) |
48208-26-0 | |
Yellow | |
MFCD08705332 | |
2r-2-1,3-dioxoisoindol-2-yl-3-1h-indol-3-yl propanoic acid, r-2-1,3-dioxoisoindoline-2-yl-3-1h-indole-3-yl propanoic acid | |
C1=CC=C2C(=C1)C(=O)N(C2=O)C(CC3=CNC4=CC=CC=C43)C(=O)O | |
334.331 | |
334.33 | |
Crystalline Powder |
Chemical Identifiers
48208-26-0 | |
334.331 | |
HPTXLHAHLXOAKV-MRXNPFEDSA-N | |
702560 | |
C1=CC=C2C(=C1)C(=O)N(C2=O)C(CC3=CNC4=CC=CC=C43)C(=O)O |
C19H14N2O4 | |
MFCD08705332 | |
2r-2-1,3-dioxoisoindol-2-yl-3-1h-indol-3-yl propanoic acid, r-2-1,3-dioxoisoindoline-2-yl-3-1h-indole-3-yl propanoic acid | |
(2R)-2-(1,3-dioxoisoindol-2-yl)-3-(1H-indol-3-yl)propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Freezer